Order Entry
Export
Orders LinkContactUsLinkComponent
Melamine 99%

Melamine 99%

Bottle
Catalog # APOSOR1013724-500G
Health
New Product
Supplier:   Apollo Scientific
Melamine 99%
Catalog # APOSOR1013724-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  108-78-1
Melamine 99%

Bottle

CAS Number:  108-78-1
Price
57,90 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  2,4,6-Triamino-s-triazine, 1,3,5-Triazine-2,4,6-triamine, 2,4,6-Triamino-1,3,5-triazine
  • CAS Number:108-78-1
  • Formula:C₃H₆N₆
  • MDL Number:MFCD00006055
  • Molecular Weight:126,12 g/mol
  • EINECS:203-615-4
Specifications

Specifications

  • Pack type:
    Bottle

Specifications