Order Entry
Export
Orders LinkContactUsLinkComponent
Phenolphthalein indicator

Phenolphthalein indicator

Bottle
Catalog # APOSOR1014628-100G
New Product
Supplier:   Apollo Scientific
Phenolphthalein indicator
Catalog # APOSOR1014628-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  77-09-8
Phenolphthalein indicator

Bottle

CAS Number:  77-09-8
Price
24,60 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
  • CAS Number:77-09-8
  • Formula:C₂₀H₁₄O₄
  • MDL Number:MFCD00005913
  • Molecular Weight:318,32 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications