Order Entry
Export
Orders LinkContactUsLinkComponent
(S)-10-Hydroxycamptothecin 98%

(S)-10-Hydroxycamptothecin 98%

Bottle
Catalog # APOSOR1008613-100G
New Product
Supplier:   Apollo Scientific
(S)-10-Hydroxycamptothecin 98%
Catalog # APOSOR1008613-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  19685-09-7
(S)-10-Hydroxycamptothecin 98%

Bottle

CAS Number:  19685-09-7
Price
1 750,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  1-HCPT, (S)-4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 10-Hydroxycamptothecin
  • CAS Number:19685-09-7
  • Formula:C₂₀H₁₆N₂O₅
  • MDL Number:MFCD02093100
  • Molecular Weight:364,35 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications