Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
DL-Malic acid
BP0000726352
Malic acid
Print
Malic acid
Bottle
Catalog # APOSOR1013709-25KG
New Product
Supplier:
Apollo Scientific
Malic acid
Catalog # APOSOR1013709-25KG
Supplier:
Apollo Scientific
New Product
CAS Number:
Expand image
Return to Top
Malic acid
Bottle
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
2 540,00 €
Log in
for availability
Unit of Measure
1 * 25 kg
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Specifications
Pack type:
Bottle
Specifications