Order Entry
Export
Orders LinkContactUsLinkComponent
Mandelic acid 99%

Mandelic acid 99%

Bottle
Catalog # APOSOR1013716-100G
New Product
Supplier:   Apollo Scientific
Mandelic acid 99%
Catalog # APOSOR1013716-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  90-64-2
Mandelic acid 99%

Bottle

CAS Number:  90-64-2
Price
24,60 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid
  • CAS Number:90-64-2
  • Formula:C₈H₈O₃
  • MDL Number:MFCD15143969
  • Molecular Weight:152,15 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications