Order Entry
Export
Orders LinkContactUsLinkComponent
 

Linalool 98%

Bottle
Catalog # APOSOR1027965-25ML
Irritant
New Product
Supplier:   Apollo Scientific
Linalool 98%
Catalog # APOSOR1027965-25ML
Supplier:  Apollo Scientific
New Product
CAS Number:  78-70-6
Linalool 98%

Bottle

CAS Number:  78-70-6
Price
21,80 €
Log in for availability
1 * 25 mL
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
  • CAS Number:78-70-6
  • Formula:C₁₀H₁₈O
  • Molecular Weight:154,25 g/mol
  • EINECS:201-134-4
Specifications

Specifications

  • Pack type:
    Bottle

Specifications