Order Entry
Export
Orders LinkContactUsLinkComponent
Propylsulphamide 98%

Propylsulphamide 98%

Bottle
Catalog # APOSOR1014847-5G
New Product
Supplier:   Apollo Scientific
Propylsulphamide 98%
Catalog # APOSOR1014847-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  147962-41-2
Propylsulphamide 98%

Bottle

CAS Number:  147962-41-2
Price
36,20 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:147962-41-2
  • Formula:C₃H₁₀N₂O₂S
  • MDL Number:MFCD12755184
  • Molecular Weight:138,19 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications