Order Entry
Export
Orders LinkContactUsLinkComponent
 

Arsenazo III indicator

Bottle
Catalog # APOSOR98227-250MG
New Product
Supplier:   Apollo Scientific
Arsenazo III indicator
Catalog # APOSOR98227-250MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1668-00-4
Arsenazo III indicator

Bottle

CAS Number:  1668-00-4
Price
21,80 €
Log in for availability
1 * 250 mg
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  2,7-Bis(2-arsonophenylazo)chromotropic acid, 2,7-Bis(2-arsonophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic acid, 2,2′-(1,8-Dihydroxy-3,6-disulphonaphthylene-2,7-bisazo)bisbenzenearsonic acid, Arsenazo Ⅲ
  • CAS Number:1668-00-4
  • Formula:C₂₂H₁₈As₂N₄O₁₄S₂
  • MDL Number:MFCD00036695
  • Molecular Weight:776,374 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications