Order Entry
Export
Orders LinkContactUsLinkComponent
Trypan Blue 80%

Trypan Blue 80%

Bottle
Catalog # APOSOR1015400-25G
New Product
Supplier:   Apollo Scientific
Trypan Blue 80%
Catalog # APOSOR1015400-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  72-57-1
Trypan Blue 80%

Bottle

CAS Number:  72-57-1
Price
46,40 €
Log in for availability
1 * 25 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
  • CAS Number:72-57-1
  • Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
  • MDL Number:MFCD00003969
  • Molecular Weight:960,81 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications