0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Propidium iodide
BP0000728772
Propidium Iodide
Bottle
APOSBUP11182-100MG
New Product
:
Apollo Scientific
Propidium Iodide
APOSBUP11182-100MG
:
Apollo Scientific
New Product
:
Propidium Iodide
Bottle
:
168,00 €
1 * 100 mg
-
+
Pack type:
Bottle