Order Entry
Export
Orders LinkContactUsLinkComponent
HCV-IN-30 ≥98%

HCV-IN-30 ≥98%

Bottle
Catalog # APOSBUP14081-50MG
New Product
Supplier:   Apollo Scientific
HCV-IN-30 ≥98%
Catalog # APOSBUP14081-50MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1007882-23-6
HCV-IN-30 ≥98%

Bottle

CAS Number:  1007882-23-6
Price
73,80 €
Log in for availability
1 * 50 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  HCV-IN-30
  • CAS Number:1007882-23-6
  • Formula:C₃₆H₄₄N₆O₄
  • Molecular Weight:624,77 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications