0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Nile red
BP0000733009
Nile Red
Bottle
APOSBUP14057-50MG
New Product
:
Apollo Scientific
Nile Red
APOSBUP14057-50MG
:
Apollo Scientific
New Product
:
Nile Red
Bottle
:
68,10 €
1 * 50 mg
-
+
Pack type:
Bottle