Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ned 19 ≥98%

Bottle
  APOSBUP12506-25MG
New Product
Ned 19 ≥98%
  APOSBUP12506-25MG
New Product
 :  874374-25-1
Ned 19 ≥98%

Bottle

 :  874374-25-1
 
458,00 €
Log in for availability
1 * 25 mg
 

Usage: Biochemical/life science applications

Synonyms:  Ned 19
  • CAS Number:874374-25-1
  • Formula:C₃₀H₃₁FN₄O₃
  • Molecular Weight:514,59 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications