Order Entry
Export
Orders LinkContactUsLinkComponent
 

AP 18 ≥90%

Bottle
Catalog # APOSBUP12981-25MG
New Product
Supplier:   Apollo Scientific
AP 18 ≥90%
Catalog # APOSBUP12981-25MG
Supplier:  Apollo Scientific
New Product
CAS Number:  55224-94-7
AP 18 ≥90%

Bottle

CAS Number:  55224-94-7
Price
183,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  4-(4-Chlorophenyl)-3-methylbut-3-en-2-oxime.
  • CAS Number:55224-94-7
  • Formula:C₁₁H₁₂ClNO
  • Molecular Weight:209,67 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications