0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
N-Cycloheptyl-2-(4,5-dihydro-1H-imidazol-2-ylsulphanyl)acetamide hydrochloride
BP0000704612
ICCB-19 hydrochloride
Bottle
APOSBUP15604-50MG
New Product
:
Apollo Scientific
ICCB-19 hydrochloride
APOSBUP15604-50MG
:
Apollo Scientific
New Product
:
ICCB-19 hydrochloride
Bottle
:
189,00 €
1 * 50 mg
-
+
Pack type:
Bottle