0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,9-Pyrazoloanthrone
BP0000698970
SP600125
Bottle
APOSBUP14499-1G
New Product
:
Apollo Scientific
SP600125
APOSBUP14499-1G
:
Apollo Scientific
New Product
:
SP600125
Bottle
:
255,00 €
1 * 1 g
-
+
Pack type:
Bottle