Order Entry
Export
Orders LinkContactUsLinkComponent
ICCB-19 hydrochloride ≥98%

ICCB-19 hydrochloride ≥98%

Bottle
Catalog # APOSBUP15604-200MG
New Product
Supplier:   Apollo Scientific
ICCB-19 hydrochloride ≥98%
Catalog # APOSBUP15604-200MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1803605-68-6
ICCB-19 hydrochloride ≥98%

Bottle

CAS Number:  1803605-68-6
Price
419,00 €
Log in for availability
1 * 200 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  ICCB-19 HCl
  • CAS Number:1803605-68-6
  • Formula:C₁₂H₂₂ClN₃OS
  • Molecular Weight:291,84 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications