MDK83190 ≥95%
Bottle
Catalog # APOSBUP13172-100MG
New Product
Supplier: Apollo Scientific
MDK83190 ≥95%
Catalog # APOSBUP13172-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 79183-19-0
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: 1-[(3,4-Dichlorophenyl)methyl]-1H-indole-2,3-dione, 1-(3,4-Dichlorobenzyl)indoline-2,3-dione
- CAS Number:79183-19-0
- Formula:C₁₅H₉Cl₂NO₂
- Molecular Weight:306,14 g/mol
Specifications
Specifications
- Pack type:Bottle



