Order Entry
Export
Orders LinkContactUsLinkComponent
MDK83190 ≥95%

MDK83190 ≥95%

Bottle
Catalog # APOSBUP13172-25MG
New Product
Supplier:   Apollo Scientific
MDK83190 ≥95%
Catalog # APOSBUP13172-25MG
Supplier:  Apollo Scientific
New Product
CAS Number:  79183-19-0
MDK83190 ≥95%

Bottle

CAS Number:  79183-19-0
Price
144,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  1-[(3,4-Dichlorophenyl)methyl]-1H-indole-2,3-dione, 1-(3,4-Dichlorobenzyl)indoline-2,3-dione
  • CAS Number:79183-19-0
  • Formula:C₁₅H₉Cl₂NO₂
  • Molecular Weight:306,14 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications