0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
NCH 51
BP0000704323
PTACH
Bottle
APOSBUP12421-200MG
New Product
:
Apollo Scientific
PTACH
APOSBUP12421-200MG
:
Apollo Scientific
New Product
:
PTACH
Bottle
:
1 130,00 €
1 * 200 mg
-
+
Pack type:
Bottle