Order Entry
Export
Orders LinkContactUsLinkComponent
Estradiol ≥98%

Estradiol ≥98%

Bottle
Catalog # APOSBUP14085-5G
New Product
Supplier:   Apollo Scientific
Estradiol ≥98%
Catalog # APOSBUP14085-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  50-28-2
Estradiol ≥98%

Bottle

CAS Number:  50-28-2
Price
300,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  beta-Estradiol, 17β-Estradiol
  • CAS Number:50-28-2
  • Formula:C₁₈H₂₄O₂
  • Molecular Weight:272,38 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications