Order Entry
Export
Orders LinkContactUsLinkComponent
MIND4-19 ≥98%

MIND4-19 ≥98%

Bottle
Catalog # APOSBUP10873-25MG
New Product
Supplier:   Apollo Scientific
MIND4-19 ≥98%
Catalog # APOSBUP10873-25MG
Supplier:  Apollo Scientific
New Product
CAS Number:  129544-85-0
MIND4-19 ≥98%

Bottle

CAS Number:  129544-85-0
Price
176,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  MIND4-19
  • CAS Number:129544-85-0
  • Formula:C₂₂H₁₉N₃OS
  • Molecular Weight:373,47 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications