Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Acid violet 43
BP0000735378
Acid violet 43
Print
Acid violet 43
Catalog # APOSOR1028904-500G
New Product
Supplier:
Apollo Scientific
Acid violet 43
Catalog # APOSOR1028904-500G
Supplier:
Apollo Scientific
New Product
CAS Number:
Expand image
Return to Top
Acid violet 43
Add to cart
CAS Number:
Price
1 540,00 €
Log in
for availability
Unit of Measure
1 * 500 g
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents