0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Acid violet 43
BP0000735378
Acid violet 43
Bottle
APOSOR1028904-25G
New Product
:
Apollo Scientific
Acid violet 43
APOSOR1028904-25G
:
Apollo Scientific
New Product
:
Acid violet 43
Bottle
:
94,10 €
1 * 25 g
-
+
Pack type:
Bottle