Order Entry
Export
Orders LinkContactUsLinkComponent
 

Acid violet 43 biological stain

Bottle
Catalog # APOSOR1028904-1G
New Product
Supplier:   Apollo Scientific
Acid violet 43 biological stain
Catalog # APOSOR1028904-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  4430-18-6
Acid violet 43 biological stain

Bottle

CAS Number:  4430-18-6
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Acid Violet 43
  • CAS Number:4430-18-6
  • Formula:C₂₁H₁₄NNaO₆S
  • Molecular Weight:431,39 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications