0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Acid violet 43
BP0000735378
Acid violet 43
APOSOR1028904-100G
New Product
:
Apollo Scientific
Acid violet 43
APOSOR1028904-100G
:
Apollo Scientific
New Product
:
Acid violet 43
:
335,00 €
1 * 100 g
-
+