Lanosterol 60%
Bottle
Catalog # APOSOR1028154-25G
New Product
Supplier: Apollo Scientific
Lanosterol 60%
Catalog # APOSOR1028154-25G
Supplier: Apollo Scientific
New Product
CAS Number: 79-63-0
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: (3S,5R,10S,13R,14R,17R)-4,4,10,13,14-Pentamethyl-17-((R)-6-methylhept-5-en-2-yl)-2,3,4,5,6,7,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol, Cryptosterol
- CAS Number:79-63-0
- Formula:C₃₀H₅₀O
- Molecular Weight:426,72 g/mol
Specifications
Specifications
- Pack type:Bottle



