Order Entry
Export
Orders LinkContactUsLinkComponent
 

Lanosterol 60%

Bottle
Catalog # APOSOR1028154-500G
New Product
Supplier:   Apollo Scientific
Lanosterol 60%
Catalog # APOSOR1028154-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  79-63-0
Lanosterol 60%

Bottle

CAS Number:  79-63-0
Price
1 130,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  (3S,5R,10S,13R,14R,17R)-4,4,10,13,14-Pentamethyl-17-((R)-6-methylhept-5-en-2-yl)-2,3,4,5,6,7,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol, Cryptosterol
  • CAS Number:79-63-0
  • Formula:C₃₀H₅₀O
  • Molecular Weight:426,72 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications