Order Entry
Export
Orders LinkContactUsLinkComponent
Calmagite indicator

Calmagite indicator

Bottle
Catalog # APOSOR1028066-25G
New Product
Supplier:   Apollo Scientific
Calmagite indicator
Catalog # APOSOR1028066-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  3147-14-6
Calmagite indicator

Bottle

CAS Number:  3147-14-6
Price
55,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:3147-14-6
  • Formula:C₁₇H₁₄N₂O₅S
  • Molecular Weight:358,37 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications