Order Entry
Export
Orders LinkContactUsLinkComponent
3-Methylcatechol ≥90%

3-Methylcatechol ≥90%

Bottle
Catalog # APOSBUP18400-500MG
New Product
Supplier:   Apollo Scientific
3-Methylcatechol ≥90%
Catalog # APOSBUP18400-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  488-17-5
3-Methylcatechol ≥90%

Bottle

CAS Number:  488-17-5
Price
40,60 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  3-Methylcatechol, 3-Methylbenzene-1,2-diol
  • CAS Number:488-17-5
  • Formula:C₇H₈O₂
  • Molecular Weight:124,14 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications