0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
20-Hydroxyecdysone
BP0000700873
Ecdysterone
Bottle
APOSOR1026667-250M
New Product
:
Apollo Scientific
Ecdysterone
APOSOR1026667-250M
:
Apollo Scientific
New Product
:
Ecdysterone
Bottle
:
34,80 €
1 * 250 mg
-
+
Pack type:
Bottle