Quinoline-4,7-diol
Bottle
Catalog # APOSOR302275-250MG
New Product
Supplier: Apollo Scientific
Quinoline-4,7-diol
Catalog # APOSOR302275-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 955938-89-3
Jump to:
Product Details & Documents
- CAS Number:955938-89-3
- Formula:C₉H₇NO₂
- MDL Number:MFCD09260461
- Molecular Weight:161.157 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



