Order Entry
Export
Orders LinkContactUsLinkComponent
 

3-(4-Bromophenyl)propionic acid

Bottle
Catalog # APOSOR3085-1G
New Product
Supplier:   Apollo Scientific
3-(4-Bromophenyl)propionic acid
Catalog # APOSOR3085-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  1643-30-7
3-(4-Bromophenyl)propionic acid

Bottle

CAS Number:  1643-30-7
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:1643-30-7
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD01310793
  • Molecular Weight:229.07 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications