Order Entry
Export
Orders LinkContactUsLinkComponent
 

Quinoline-4,7-diol

Bottle
Catalog # APOSOR302275-25G
New Product
Supplier:   Apollo Scientific
Quinoline-4,7-diol
Catalog # APOSOR302275-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  955938-89-3
Quinoline-4,7-diol

Bottle

CAS Number:  955938-89-3
Price
801,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:955938-89-3
  • Formula:C₉H₇NO₂
  • MDL Number:MFCD09260461
  • Molecular Weight:161.157 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications