Order Entry
Export
Orders LinkContactUsLinkComponent
 

5-Nitroisoquinoline

Bottle
Catalog # APOSOR21601-5G
New Product
Supplier:   Apollo Scientific
5-Nitroisoquinoline
Catalog # APOSOR21601-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  607-32-9
5-Nitroisoquinoline

Bottle

CAS Number:  607-32-9
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:607-32-9
  • Formula:C₉H₆N₂O₂
  • MDL Number:MFCD00006905
  • Molecular Weight:174.16 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications