Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,3'-Bipyridine 95%

Bottle
Catalog # APOSOR931059-500G
New Product
Supplier:   Apollo Scientific
2,3'-Bipyridine 95%
Catalog # APOSOR931059-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  581-50-0
2,3'-Bipyridine 95%

Bottle

CAS Number:  581-50-0
Price
2 150,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:581-50-0
  • Formula:C₁₀H₈N₂
  • MDL Number:MFCD00006215
  • Molecular Weight:156.184 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications