Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Dithiane 98%

1,3-Dithiane 98%

Bottle
Catalog # APOSOR51910-500G
New Product
Supplier:   Apollo Scientific
1,3-Dithiane 98%
Catalog # APOSOR51910-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  505-23-7
1,3-Dithiane 98%

Bottle

CAS Number:  505-23-7
Price
915,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:505-23-7
  • Formula:C₄H₈S₂
  • MDL Number:MFCD00006654
  • Molecular Weight:120.24 g/mol
  • Boiling Pt:207…208 °C
  • Melting Pt:52…54 °C
  • Flash Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:208-006-7
  • UN:3335
  • ADR:9,III
  •  :Density: 1.36 g/cm³ (25 °C)
Specifications

Specifications

  • Pack type:
    Bottle

Specifications