4-(2-Hydroxyethoxy)benzaldehyde 95%
Bottle
Catalog # APOSOR922883-1G
New Product
Supplier: Apollo Scientific
4-(2-Hydroxyethoxy)benzaldehyde 95%
Catalog # APOSOR922883-1G
Supplier: Apollo Scientific
New Product
CAS Number: 22042-73-5
Jump to:
Product Details & Documents
- CAS Number:22042-73-5
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00191450
- Molecular Weight:166.1739 g/mol
- Melting Pt:40 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



