Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Dibromopyridine

2,6-Dibromopyridine

Bottle
Catalog # APOSOR28767-500G
Toxic
New Product
Supplier:   Apollo Scientific
2,6-Dibromopyridine
Catalog # APOSOR28767-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  626-05-1
2,6-Dibromopyridine

Bottle

CAS Number:  626-05-1
Price
145,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:626-05-1
  • Formula:C₅H₃Br₂N
  • MDL Number:MFCD00006223
  • Molecular Weight:236.89 g/mol
  • Boiling Pt:255 °C
  • Melting Pt:118…119 °C
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:210-926-9
  • UN:2811
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Bottle

Specifications