1,3-Benzenedithiol
Bottle
Catalog # APOSOR55264-250MG
New Product
Supplier: Apollo Scientific
1,3-Benzenedithiol
Catalog # APOSOR55264-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 626-04-0
Jump to:
Product Details & Documents
Synonyms: Benzene-1,3-dithiol
- CAS Number:626-04-0
- Formula:C₆H₆S₂
- MDL Number:MFCD00004842
- Molecular Weight:142.25 g/mol
- Boiling Pt:244 °C
- Melting Pt:24…25 °C
- Density:1.24 g/cm³ (25 °C)
- Flash Pt:113 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



