Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Propanediol

1,3-Propanediol

Bottle
Catalog # APOSOR0974-500G
New Product
Supplier:   Apollo Scientific
1,3-Propanediol
Catalog # APOSOR0974-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  504-63-2
1,3-Propanediol

Bottle

CAS Number:  504-63-2
Price
57,90 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  1,3-Propylene glycol, 1,3-Dihydroxy propane
  • CAS Number:504-63-2
  • Formula:HO(CH₂)₃OH
  • MDL Number:MFCD00002949
  • Molecular Weight:76.1 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications