Order Entry
Export
Orders LinkContactUsLinkComponent
1-Octanol 98%

1-Octanol 98%

Bottle
Catalog # APOSOR40039-500ML
Irritant
New Product
Supplier:   Apollo Scientific
1-Octanol 98%
Catalog # APOSOR40039-500ML
Supplier:  Apollo Scientific
New Product
CAS Number:  111-87-5
1-Octanol 98%

Bottle

CAS Number:  111-87-5
Price
57,90 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  n-Octanol
  • CAS Number:111-87-5
  • Formula:H₃C(CH₂)₆CH₂OH
  • MDL Number:MFCD00002988
  • Molecular Weight:130.23 g/mol
  • Boiling Pt:193…195 °C
  • Melting Pt:–16 °C
  • Density:0.825 g/cm³ (25 °C)
  • Flash Pt:81 °C
  • Storage Temperature:Ambient
  • EINECS:203-917-6
Specifications

Specifications

  • Pack type:
    Bottle

Specifications