3',4'-Dihydroxyacetophenone 98%
Bottle
Catalog # APOSOR345013-500G
New Product
Supplier: Apollo Scientific
3',4'-Dihydroxyacetophenone 98%
Catalog # APOSOR345013-500G
Supplier: Apollo Scientific
New Product
CAS Number: 1197-09-7
Jump to:
Product Details & Documents
Synonyms: 1-(3,4-Dihydroxyphenyl)ethan-1-one, Acetopyrocatechol
- CAS Number:1197-09-7
- Formula:C₈H₈O₃
- MDL Number:MFCD00016439
- Molecular Weight:152.15 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



