Order Entry
Export
Orders LinkContactUsLinkComponent
3',4'-Dihydroxyacetophenone 98%

3',4'-Dihydroxyacetophenone 98%

Bottle
Catalog # APOSOR345013-5G
New Product
Supplier:   Apollo Scientific
3',4'-Dihydroxyacetophenone 98%
Catalog # APOSOR345013-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  1197-09-7
3',4'-Dihydroxyacetophenone 98%

Bottle

CAS Number:  1197-09-7
Price
24,60 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  1-(3,4-Dihydroxyphenyl)ethan-1-one, Acetopyrocatechol
  • CAS Number:1197-09-7
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00016439
  • Molecular Weight:152.15 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Cat. No.:
    APOSOR345013-5G

Specifications