Order Entry
Export
Orders LinkContactUsLinkComponent
1,1-Diphenylethylene 98%

1,1-Diphenylethylene 98%

Bottle
Catalog # APOSOR315713-2.5KG
New Product
Supplier:   Apollo Scientific
1,1-Diphenylethylene 98%
Catalog # APOSOR315713-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  530-48-3
1,1-Diphenylethylene 98%

Bottle

CAS Number:  530-48-3
Price
2 230,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene
  • CAS Number:530-48-3
  • Formula:C₁₄H₁₂
  • MDL Number:MFCD00008583
  • Molecular Weight:180,24508 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications