1,1-Diphenylethylene 98%
Bottle
Catalog # APOSOR315713-5G
New Product
Supplier: Apollo Scientific
1,1-Diphenylethylene 98%
Catalog # APOSOR315713-5G
Supplier: Apollo Scientific
New Product
CAS Number: 530-48-3
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms: asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene
- CAS Number:530-48-3
- Formula:C₁₄H₁₂
- MDL Number:MFCD00008583
- Molecular Weight:180,24508 g/mol
Specifications
Specifications
- Pack type:Bottle



