4-Ethylpyrocatechol 97%
Bottle
Catalog # APOSOR925237-25G
New Product
Supplier: Apollo Scientific
4-Ethylpyrocatechol 97%
Catalog # APOSOR925237-25G
Supplier: Apollo Scientific
New Product
CAS Number: 1124-39-6
Jump to:
Product Details & Documents
Synonyms: 4-Ethylbenzene-1,2-diol, 4-Ethylcatechol
- CAS Number:1124-39-6
- Formula:C₈H₁₀O₂
- MDL Number:MFCD00015847
- Molecular Weight:138.17 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



