Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
4,4'-Dihydroxyazobenzene
4,4'-Dihydroxyazobenzene
4,4'-Dihydroxyazobenzene
4,4'-Dihydroxyazobenzene
Bottle
APOSOR305565-5G
New Product
:
Apollo Scientific
4,4'-Dihydroxyazobenzene
APOSOR305565-5G
:
Apollo Scientific
New Product
:
2050-16-0
Expand image
4,4'-Dihydroxyazobenzene
Bottle
Add to cart
:
2050-16-0
103,00 €
Unit of Measure
1 * 5 g
-
+
Add to cart
:
4,4-Dihydroxyazobenzene, 4,4'-Azodiphenol
:
2050-16-0
:
C₁₂H₁₀N₂O₂
:
MFCD00045778
:
214.22 g/mol
:
215 °C
:
Ambient
Pack type:
Bottle