Orders LinkContactUsLinkComponent
4-Hydroxycinnamic acid 98%
Bottle
  APOSOR21362-25G
New Product
4-Hydroxycinnamic acid 98%
  APOSOR21362-25G
New Product
 :  7400-08-0
4-Hydroxycinnamic acid 98%
Bottle
 :  7400-08-0
23,20 €
1 * 25 g
 :  p-Coumaric acid, p-Cumaric acid, 3-(4-Hydroxyphenyl)acrylic acid, p-Hydroxy-Cinnamic Acid
  •  :7400-08-0
  •  :C₉H₈O₃
  •  :MFCD00004399
  •  :164.16 g/mol
  •  :346 °C
  •  :212…214 °C
  •  :Ambient
  • Pack type:
    Bottle