Order Entry
Export
Orders LinkContactUsLinkComponent
2,2-Dichloroacetamide

2,2-Dichloroacetamide

Bottle
Catalog # APOSOR14320-100G
Irritant
New Product
Supplier:   Apollo Scientific
2,2-Dichloroacetamide
Catalog # APOSOR14320-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  683-72-7
2,2-Dichloroacetamide

Bottle

CAS Number:  683-72-7
Price
119,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:683-72-7
  • Formula:C₂H₃Cl₂NO
  • MDL Number:MFCD00008015
  • Molecular Weight:127.96 g/mol
  • Boiling Pt:233…234 °C
  • Melting Pt:96…101 °C
  • Storage Temperature:Ambient
  • EINECS:211-674-2
Specifications

Specifications

  • Pack type:
    Bottle

Specifications